missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Bromoadenosine, 98%, ACROS Organics™

Manufacturer:  Acros Organics AC155782500

 View more versions of this product

Catalog No. AC155782500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

8-Bromoadenosine
Glass bottle
-53° to -56° (20°C, 589nm) (c=2.5, DMSO)
250mg
97.5
98%
MFCD00005733
VJUPMOPLUQHMLE-UUOKFMHZSA-N
(2R,3R,4S,5R)-2-(6-amino-8-bromopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
96544
346.13
≥97.5%
Authentic
−54.50° (20°C c=2.5,DMSO)
White to Cream
2946-39-6
100.0
C10H12BrN5O4
8-bromoadenosine, bromoadenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, adenosine, 8-bromo, 6-amino-8-bromopurine riboside, 8-bromo-adenosine, 8-bromoadenine-9-beta-d-ribofuranoside, 8-bromo adenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 4s,2r,3r,5r-2-6-amino-8-bromopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol
C1=NC2=C(C(=N1)N)N=C(N2C3C(C(C(O3)CO)O)O)Br
346.141
CHEBI:2312
Crystalline Powder

Chemical Identifiers

2946-39-6
346.141
VJUPMOPLUQHMLE-UUOKFMHZSA-N
96544
(2R,3R,4S,5R)-2-(6-amino-8-bromopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
C10H12BrN5O4
MFCD00005733
8-bromoadenosine, bromoadenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-9h-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, adenosine, 8-bromo, 6-amino-8-bromopurine riboside, 8-bromo-adenosine, 8-bromoadenine-9-beta-d-ribofuranoside, 8-bromo adenosine, 2r,3r,4s,5r-2-6-amino-8-bromo-purin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 4s,2r,3r,5r-2-6-amino-8-bromopurin-9-yl-5-hydroxymethyl oxolane-3,4-diol
CHEBI:2312
C1=NC2=C(C(=N1)N)N=C(N2C3C(C(C(O3)CO)O)O)Br
SDS Documents