Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Bromoadenosine, 98%

$59.96 - $59.96

Chemical Identifiers

Chemical Name or Material 8-Bromoadenosine
CAS 2946-39-6
Molecular Formula C10H12BrN5O4
Formula Weight 346.15
MDL Number MFCD00005733
Beilstein 627737
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL03544MD

  SDS
alfa aesar™
L03544MD
250mg Each for $59.96
Description & Specifications

Specifications

Formula Weight 346.15
Assay 98%
Molecular Formula C10H12BrN5O4
MDL Number MFCD00005733
CAS 2946-39-6
Melting Point ≈211°C
Beilstein 627737
Quantity 250mg
Optical Rotation −33° (c=1 in 1N HCl)
Recommended Storage 2° to 8°C
Chemical Name or Material 8-Bromoadenosine

Applications
8-Bromoadenosine is an adenosine binding inhibitor.

Solubility
Soluble in DMSO, and DMF. Insoluble in water.

Notes
Air sensitive. Store under inert gas. Store away from oxidizing agents, air.

Safety and Handling

2° to 8°C