missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Bromoadenosine-3',5'-cyclic monophosphate

Manufacturer:  Alfa Aesar™ J64469MC

 View more versions of this product

Catalog No. AAJ64469MC



Specifications

Specifications

8-Bromoadenosine-3',5'-cyclic monophosphate
Brown
100mg
C10H11BrN5O6P
8-br-camp, 8-bromo cyclic adenosine monophosphate, 8-bromo-camp, 8-bromo-cyclic 3',5'-amp, 8-bromo-cyclic amp, 8-bromoadenosine 3',5'-cyclic monophosphate, 8-bromoadenosine 3',5'-monophosphate, 8-bromoadenosine cyclic 3',5'-phosphate, bcamp, cyclic 8-bromoadenosine 3',5'-monophosphate
C1C2C(C(C(O2)N3C4=C(C(=NC=N4)N)N=C3Br)O)OP(=O)(O1)O
408.105
CHEBI:64211
Powder
Hygroscopic
254°C
23583-48-4
MFCD00075580
DVKQVRZMKBDMDH-UUOKFMHZSA-N
(4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
32014
408.11

Chemical Identifiers

23583-48-4
408.105
DVKQVRZMKBDMDH-UUOKFMHZSA-N
32014
(4aR,6R,7R,7aS)-6-(6-amino-8-bromopurin-9-yl)-2-hydroxy-2-oxo-4a,6,7,7a-tetrahydro-4H-furo[3,2-d][1,3,2]dioxaphosphinin-7-ol
C10H11BrN5O6P
MFCD00075580
8-br-camp, 8-bromo cyclic adenosine monophosphate, 8-bromo-camp, 8-bromo-cyclic 3',5'-amp, 8-bromo-cyclic amp, 8-bromoadenosine 3',5'-cyclic monophosphate, 8-bromoadenosine 3',5'-monophosphate, 8-bromoadenosine cyclic 3',5'-phosphate, bcamp, cyclic 8-bromoadenosine 3',5'-monophosphate
CHEBI:64211
C1C2C(C(C(O2)N3C4=C(C(=NC=N4)N)N=C3Br)O)OP(=O)(O1)O
Safety and Handling

Safety and Handling

EINECSNumber : 245-760-6

TSCA : No

Recommended Storage : Store at -20°C

Documents