8-Bromoadenine, Fisher BioReagents
Manufacturer:  Fisher Scientific Company BP2507100
 View more versions of this product
Catalog No. BP2507100
Specifications
Description
8-Bromoadenine; Light Tan Powder; Fisher BioReagents; 100mg; Amber Glass; F.W. 214.03; CHNBr; CAS: 6974-78-3
  • Adenine analog in which the hydrogen in the 8-position is replaced by a bromine atom
  • Useful for the study of metabolic processes
  • Intended for biochemical research applications 
Description & Specifications
Specifications
214.03
100mg
Pass Test
6974-78-3
Amber Glass
Powder/Solid
White
8-Bromoadenine
6-Amino-8-bromopurine
C5H4N5Br
Chemical Identifiers
6974-78-3
C5H4N5Br
6-Amino-8-bromopurine
Powder/Solid
214.03
8-Bromoadenine
Safety and Handling
SDS & Safety Documents
Recommended Storage : Store below 0° C, desiccate