Please login to your online account to display your discounted pricing

8-Bromoadenine, Fisher BioReagents

$113.83 - $113.83

Chemical Identifiers

Chemical Name or Material 8-Bromoadenine
CAS 6974-78-3
Physical Form Powder/Solid
Molecular Formula C5H4N5Br
Formula Weight 214.03
Synonym 6-Amino-8-bromopurine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP2507100

  SDS
fisher scientific company
BP2507100
Amber Glass 100mg Each for $113.83
Case of 5 Each for $569.15
Description & Specifications

Specifications

Physical Form Powder/Solid
Formula Weight 214.03
Quantity 100mg
Color White
Chemical Name or Material 8-Bromoadenine
ChemAlert Storage Symbol Gray
Recommended Storage Store below 0° C, desiccate
Identification Pass Test
Packaging Amber Glass
Molecular Formula C5H4N5Br
CAS 6974-78-3
Synonym 6-Amino-8-bromopurine

8-Bromoadenine is used in several biochemical research applications.

Safety and Handling
 

Store below 0° C, desiccate