8-Bromo-cAMP, sodium salt, Tocris
Cell-permeable cAMP analog
Manufacturer:  R & D Systems 114010
 View more versions of this product
Catalog No. 11-401-0
Specifications Description & Specifications
Specifications
8-Bromo-cAMP, sodium salt
>99%
430.09
10mg
76939-46-3
C10H10BrN5NaO6P
8-Bromoadenosine-3', 5'-cyclic monophosphate sodium salt
Chemical Identifiers
>99%
430.09
8-Bromo-cAMP, sodium salt
C10H10BrN5NaO6P
8-Bromoadenosine-3', 5'-cyclic monophosphate sodium salt
Safety and Handling
Recommended Storage : Desiccate at -20°C