Please login to your online account to display your discounted pricing

8-Bromo-cAMP, sodium salt, Tocris

Cell-permeable cAMP analog

$89.12 - $316.20

Chemical Identifiers

Chemical Name or Material 8-Bromo-cAMP, sodium salt
Assay Percent Range >99%
Molecular Formula C10H10BrN5NaO6P
Formula Weight 430.09
Synonym 8-Bromoadenosine-3', 5'-cyclic monophosphate sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

11-405-0

 
r & d systems
114050*
50mg Each for $316.20

11-401-0

 
r & d systems
114010
10mg Each for $89.12
Description & Specifications

Specifications

Formula Weight 430.09
Quantity 50mg
Chemical Name or Material 8-Bromo-cAMP, sodium salt
Assay Percent Range >99%
Molecular Formula C10H10BrN5NaO6P
CAS 76939-46-3
Synonym 8-Bromoadenosine-3', 5'-cyclic monophosphate sodium salt
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C