Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Bromo-1,6-naphthyridin-2(1H)-one, 98%

$100.51 - $344.80

Chemical Identifiers

Chemical Name or Material 8-Bromo-1,6-naphthyridin-2(1H)-one
CAS 902837-41-6
Molecular Formula C8H5BrN2O
Formula Weight 225.05
MDL Number MFCD08060938
Synonym 8-Bromo-2-hydroxy-1,6-naphthyridine; 6-Bromo-1,6-naphthyridin-2-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5004503

 
alfa aesar™
H5004503
1g Each for $344.80

AAH50045MD

 
alfa aesar™
H50045MD
250mg Each for $100.51
Description & Specifications

Specifications

Formula Weight 225.05
Molecular Formula C8H5BrN2O
Quantity 1g
Chemical Name or Material 8-Bromo-1,6-naphthyridin-2(1H)-one
Assay 98%
MDL Number MFCD08060938
CAS 902837-41-6
Synonym 8-Bromo-2-hydroxy-1,6-naphthyridine; 6-Bromo-1,6-naphthyridin-2-ol

Applications
8-Bromo-1,6-naphthyridin-2(1H)-one is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.