missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438880010
Specifications
8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene | |
Glass Bottle | |
1g | |
94.0 | |
94% min. (HPLC) | |
8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-3-en-3-yl trifluoromethanesulfonate, trifluoro-methanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester, trifluoro-methanesulfonic acid 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl ester, trifluoromethanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester | |
C1CC2C=C(CC1N2CC3=CC=CC=C3)OS(=O)(=O)C(F)(F)F | |
347.36 | |
347.36 | |
95% |
Authentic | |
Orange | |
949902-02-7 | |
100.0 | |
C15H16F3NO3S | |
GBUXWXWPNDHMRG-UHFFFAOYSA-N | |
(8-benzyl-8-azabicyclo[3.2.1]oct-3-en-3-yl) trifluoromethanesulfonate | |
49835910 | |
Viscous Liquid, May Contain Suspension |
Chemical Identifiers
949902-02-7 | |
347.36 | |
8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-3-en-3-yl trifluoromethanesulfonate, trifluoro-methanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester, trifluoro-methanesulfonic acid 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl ester, trifluoromethanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester | |
(8-benzyl-8-azabicyclo[3.2.1]oct-3-en-3-yl) trifluoromethanesulfonate |
C15H16F3NO3S | |
GBUXWXWPNDHMRG-UHFFFAOYSA-N | |
49835910 | |
C1CC2C=C(CC1N2CC3=CC=CC=C3)OS(=O)(=O)C(F)(F)F |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im
GHS Signal Word: Danger