Please login to your online account to display your discounted pricing

8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene
CAS 949902-02-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94.0%
Assay Percent Range 95%
Molecular Formula C15H16F3NO3S
Formula Weight 347.36
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC438880050

 
thermo fisher scientific
AC438880050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC438880010

 
thermo fisher scientific
AC438880010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Viscous Liquid, May Contain Suspension
Assay ≥94.0%
Formula Weight 347.36
Quantity 5g
Color Dark orange
CAS Max % 100.0
CAS Min % 94.0
Infrared Spectrum Authentic
Molecular Formula C15H16F3NO3S
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene
Assay Percent Range 95%
CAS 949902-02-7
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.