missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438880050

 View more versions of this product

Catalog No. AC438880050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

8-Benzyl-3-(trifluoromethylsulfonyloxy)-8-azabicyclo[3.2.1]oct-3-ene
Orange
949902-02-7
100.0
C15H16F3NO3S
GBUXWXWPNDHMRG-UHFFFAOYSA-N
(8-benzyl-8-azabicyclo[3.2.1]oct-3-en-3-yl) trifluoromethanesulfonate
49835910
Viscous Liquid, May Contain Suspension
Authentic
5g
94.0
94% min. (HPLC)
8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-3-en-3-yl trifluoromethanesulfonate, trifluoro-methanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester, trifluoro-methanesulfonic acid 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl ester, trifluoromethanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester
C1CC2C=C(CC1N2CC3=CC=CC=C3)OS(=O)(=O)C(F)(F)F
347.36
347.36
95%

Chemical Identifiers

949902-02-7
347.36
8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl trifluoromethanesulfonate, 8-benzyl-8-azabicyclo 3.2.1 oct-3-en-3-yl trifluoromethanesulfonate, trifluoro-methanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester, trifluoro-methanesulfonic acid 8-benzyl-8-azabicyclo 3.2.1 oct-2-en-3-yl ester, trifluoromethanesulfonic acid 8-benzyl-8-aza-bicyclo 3.2.1 oct-2-en-3-yl ester
(8-benzyl-8-azabicyclo[3.2.1]oct-3-en-3-yl) trifluoromethanesulfonate
C15H16F3NO3S
GBUXWXWPNDHMRG-UHFFFAOYSA-N
49835910
C1CC2C=C(CC1N2CC3=CC=CC=C3)OS(=O)(=O)C(F)(F)F
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off im

GHS Signal Word: Danger

Documents