missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Aminonaphthalene-1-sulfonic acid, 98%

$45.18 - $387.07

Chemical Identifiers

CAS 82-75-7
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00035730
InChI Key CYJJLCDCWVZEDZ-UHFFFAOYSA-N
Synonym peri acid, 8-amino-1-naphthalenesulfonic acid, 1-naphthylamine-8-sulfonic acid, 1-naphthalenesulfonic acid, 8-amino, 8-aminonaphthalene-1-sulphonic acid, naphthylaminemonosulfonic acid s, 1-amino-8-naphthalene sulfonate, 1-aminonaphthalene-8-sulfonic acid, schollkopf's acid van, schollkopf's acid
PubChem CID 6722
IUPAC Name 8-aminonaphthalene-1-sulfonic acid
SMILES C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1761614 View Documents Alfa Aesar™
L1761614
25g Each for $45.18
 
AAL1761622 View Documents Alfa Aesar™
L1761622
100g Each for $102.24
 
AAL1761636 View Documents Alfa Aesar™
L1761636
500g Each for $387.07
 
Specifications

Chemical Identifiers

82-75-7
223.246
CYJJLCDCWVZEDZ-UHFFFAOYSA-N
6722
C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O
C10H9NO3S
MFCD00035730
peri acid, 8-amino-1-naphthalenesulfonic acid, 1-naphthylamine-8-sulfonic acid, 1-naphthalenesulfonic acid, 8-amino, 8-aminonaphthalene-1-sulphonic acid, naphthylaminemonosulfonic acid s, 1-amino-8-naphthalene sulfonate, 1-aminonaphthalene-8-sulfonic acid, schollkopf's acid van, schollkopf's acid
8-aminonaphthalene-1-sulfonic acid

Specifications

>300°C
82-75-7
MFCD00035730
14,6405
CYJJLCDCWVZEDZ-UHFFFAOYSA-N
8-aminonaphthalene-1-sulfonic acid
6722
98%
25g
C10H9NO3S
983230
peri acid, 8-amino-1-naphthalenesulfonic acid, 1-naphthylamine-8-sulfonic acid, 1-naphthalenesulfonic acid, 8-amino, 8-aminonaphthalene-1-sulphonic acid, naphthylaminemonosulfonic acid s, 1-amino-8-naphthalene sulfonate, 1-aminonaphthalene-8-sulfonic acid, schollkopf's acid van, schollkopf's acid
C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O
223.246
223.25
8-Aminonaphthalene-1-sulfonic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA