Please login to your online account to display your discounted pricing

8-Aminoadenine, Tocris™

Adenine receptor agonist

$126.32 - $534.65

Chemical Identifiers

Chemical Name or Material 8-Aminoadenine
Assay Percent Range >98%
Molecular Formula C5H6N6
Formula Weight 150.14
Synonym 9H-Purine-6, 8-diamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

405010

 
r & d systems
405010
10mg Each for $126.32

405050

 
r & d systems
405050
50mg Each for $534.65
Description & Specifications

Specifications

Formula Weight 150.14
Quantity 10mg
Chemical Name or Material 8-Aminoadenine
Molecular Formula C5H6N6
Assay Percent Range >98%
Synonym 9H-Purine-6, 8-diamine
CAS 28128-33-8
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C