Please login to your online account to display your discounted pricing

8-Amino-5-bromoisoquinoline, 97%, ACROS Organics™


Quantity 1g

Manufacturer: thermo fisher scientific  AC431080010

 View more versions of this product

Catalog No. AC431080010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 223.07
Quantity 1g
Color Off-white to brown
Melting Point 186° to 190°C
CAS Min % 96.0
CAS Max % 100.0
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. Avoid breathing d
Infrared Spectrum Authentic
Assay Percent Range 97%
Chemical Name or Material 8-Amino-5-bromoisoquinoline
Molecular Formula C9H7BrN2
CAS 90721-35-0
Health Hazard 2 GHS P Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.

Specifications

Chemical Name or Material 8-Amino-5-bromoisoquinoline
CAS 90721-35-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H7BrN2
Formula Weight 223.07
Accessories

Warning

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. Avoid breathing d

GHS P Statement Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.