Please login to your online account to display your discounted pricing

8-Amino-5-bromoisoquinoline, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 8-Amino-5-bromoisoquinoline
CAS 90721-35-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H7BrN2
Formula Weight 223.07
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC431080050

  SDS
thermo fisher scientific
AC431080050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. Avoid breathing dust/fume/gas/mist/vapors/spray. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Health Hazard 2 GHS H Statement
Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled.
Formula Weight 223.07
Assay ≥96.0%
Physical Form Crystalline Powder
Quantity 5g
Color Off-white to brown
Melting Point 186° to 190°C
CAS Min % 96.0
CAS Max % 100.0
Molecular Formula C9H7BrN2
Health Hazard 1 Warning
Assay Percent Range 97%
Chemical Name or Material 8-Amino-5-bromoisoquinoline
CAS 90721-35-0
Infrared Spectrum Authentic