Please login to your online account to display your discounted pricing

8-Amino-2H-1,4-benzoxazin-3(4H)-one, 95%, Acros Organics


Quantity 1g

Manufacturer: thermo fisher scientific  AC445700010

 View more versions of this product

Catalog No. AC445700010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material 8-Amino-2H-1, 4-benzoxazin-3(4H)-one
CAS 321126-82-3
Assay Percent Range 95%
Molecular Formula C8H8N2O2
Formula Weight 164.16
Quantity 1g

Specifications

Chemical Name or Material 8-Amino-2H-1, 4-benzoxazin-3(4H)-one
Assay Percent Range 95%
Molecular Formula C8H8N2O2
Formula Weight 164.16