Please login to your online account to display your discounted pricing

8-Amino-2H-1,4-benzoxazin-3(4H)-one, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 8-Amino-2H-1, 4-benzoxazin-3(4H)-one
Assay Percent Range 95%
Molecular Formula C8H8N2O2
Formula Weight 164.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC445700010

  SDS
thermo fisher scientific
AC445700010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC445700050

  SDS
thermo fisher scientific
AC445700050
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 164.16
Quantity 1g
Chemical Name or Material 8-Amino-2H-1, 4-benzoxazin-3(4H)-one
Assay Percent Range 95%
Molecular Formula C8H8N2O2
CAS 321126-82-3