Please login to your online account to display your discounted pricing

8-Amino-2-naphthol, 94%, ACROS Organics™

$134.44 - $134.44

Chemical Identifiers

Chemical Name or Material 8-Amino-2-naphthol
CAS 118-46-7
CAS Min % 92.0
CAS Max % 100.0
Assay ≥92%
Assay Percent Range 94%
Molecular Formula C10H9NO
Formula Weight 159.18
MDL Number MFCD00004031
Synonym 1-Amino-7-naphthol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC346390050

  SDS
fisher scientific acros
346390050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC346390250

  SDS
thermo fisher scientific
346390250
Glass bottle 25g Each for $134.44
Description & Specifications

Specifications

Assay ≥92%
Physical Form Powder or Crystals
Formula Weight 159.18
Quantity 5g
Color Gray to brown
Beilstein 13, 685
CAS Min % 92.0
CAS Max % 100.0
Synonym 1-Amino-7-naphthol
Infrared Spectrum Authentic
Chemical Name or Material 8-Amino-2-naphthol
Molecular Formula C10H9NO
Assay Percent Range 94%
CAS 118-46-7
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 1 Warning
MDL Number MFCD00004031
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Safety and Handling

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.