Please login to your online account to display your discounted pricing

Alfa Aesar™ 8-Acetoxyquinoline, 97%

$168.76 - $168.76

Chemical Identifiers

Chemical Name or Material 8-Acetoxyquinoline
CAS 2598-29-0
Molecular Formula C11H9NO2
Formula Weight 187.20g/mol
MDL Number MFCD00014665
Beilstein 135053
Synonym 8-Quinolinyl acetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0302403

 
alfa aesar™
L03024-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0302406

 
alfa aesar™
L03024-06
5g Each for $168.76
Description & Specifications

Specifications

Melting Point 53° to 57°C
Quantity 1g
Odor Odorless
Chemical Name or Material 8-Acetoxyquinoline
CAS 2598-29-0
Assay 97%
Molecular Formula C11H9NO2
Formula Weight 187.20g/mol
MDL Number MFCD00014665
Beilstein 135053
Synonym 8-Quinolinyl acetate
RTECS Number VC4376000