Please login to your online account to display your discounted pricing

Alfa Aesar™ 8,11-Eicosadiynoic acid, 98%


Quantity 1mg

Manufacturer: alfa aesar™  J67017-MCR

 View more versions of this product

Catalog No. AAJ67017MCR

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Color Pale yellow
Quantity 1mg
Sensitivity Air Sensitive
Chemical Name or Material 8,11-Eicosadiynoic acid
CAS 82073-91-4
Physical Form Waxy Solid
Assay 98%
Formula Weight 304.47g/mol
MDL Number MFCD00210938
Synonym EDYA
Storage Note 1 -30° to -10°C

Specifications

Chemical Name or Material 8,11-Eicosadiynoic acid
CAS 82073-91-4
Formula Weight 304.47g/mol
MDL Number MFCD00210938
Synonym EDYA
Accessories

-30° to -10°C