Please login to your online account to display your discounted pricing

Alfa Aesar™ 8,11-Eicosadiynoic acid, 98%

Chemical Identifiers

Chemical Name or Material 8,11-Eicosadiynoic acid
CAS 82073-91-4
Formula Weight 304.47g/mol
MDL Number MFCD00210938
Synonym EDYA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67017MCR

 
alfa aesar™
J67017-MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Pale yellow
Quantity 1mg
Sensitivity Air Sensitive
Chemical Name or Material 8,11-Eicosadiynoic acid
CAS 82073-91-4
Physical Form Waxy Solid
Assay 98%
Formula Weight 304.47g/mol
MDL Number MFCD00210938
Synonym EDYA
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C