Please login to your online account to display your discounted pricing

Alfa Aesar™ 7H-Benz[de]anthracen-7-one, 99%

$72.80 - $187.80

Chemical Identifiers

Chemical Name or Material 7H-Benz[de]anthracen-7-one
CAS 82-05-3
Molecular Formula C17H10O
Formula Weight 230.27
MDL Number MFCD00003585
Beilstein 1455646
Merck Index 14,1063
Synonym Benzanthrone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2532214

 
alfa aesar™
B2532214
25g Each for $72.80

AAB2532222

 
alfa aesar™
B2532222
100g Each for $187.80
Description & Specifications

Specifications

Formula Weight 230.27
Chemical Name or Material 7H-Benz[de]anthracen-7-one
TSCA TSCA
Assay 99%
Molecular Formula C17H10O
MDL Number MFCD00003585
CAS 82-05-3
Beilstein 1455646
Merck Index 14,1063
Synonym Benzanthrone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number CX5075000
Melting Point 171° to 175°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.