Applied Biosystems™ 7500 Real-Time PCR System, desktop DFS Item
The 7500 Real-Time PCR System is a powerful platform for labs requiring superior performance and maximum dye versatility.
Specifications
orderInfo Ambient Temperature
Dynamic Range 9 logs
Precision 99.7% confidence (5000 & 10000 copies)
Sensitivity 1 copy of RNase P gene (human genomic DNA)
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
4351105 View Documents Applied Biosystems™
LS4351105
Desktop Computer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4351104 View Documents Applied Biosystems™
LS4351104
Notebook Computer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
This system is a sophisticated platform for users who require extended capabilities and maximum versatility. The 3rd generation platform features an innovative optical system that enhances sensitivity and lets you access a broader range of fluorophores. T
  • Powerful, five-color platform is calibrated for the broadest range of dyes available: FAM™⁄SYBR™ Green I, VIC™⁄JOE™, NED™⁄ TAMRA™⁄ Cy3™, ROX™⁄Texas Red™, and
    Order Info
    Ambient Temperature
Specifications
Specifications
Ambient Temperature
9 logs
99.7% confidence (5000 & 10000 copies)
1 copy of RNase P gene (human genomic DNA)