Applied Biosystems™ 7500 Fast Real-Time PCR System DFS Item
Offers maximum performance in the minimum time - fully optimized for Fast cycling, the 7500 Fast delivers high-quality results in as little as 30 minutes.
Specifications
Quantity 1 System
Item Description Throughput: 4 to 5-96-well plates per day; Scan Time: <2 hr. (Standard), 36 min. (Fast); Precision: Distinguish between 5000 to 10000 genome equivalents (two-fold copy number difference) with 99.7% confidence
Pre-Calibrated Dyes Cy3™, Cy5™, FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, Texas Red™, VIC™
Height (Metric) 49cm
Depth (Metric) 45cm
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity  
Catalog Number Mfr. No. Includes Price Quantity  
4351106 View Documents Applied Biosystems™
LS4351106
Laptop Computer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
4351107 View Documents Applied Biosystems™
LS4351107
Desktop Computer N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
We supply a Dell™ Notebook or Tower computer with the 7500 Fast System.
  • Specially designed Fast block ensures thermal uniformity at top speeds.
  • Faster ramp rates enable rapid results without compromising extension times or assay quality.
  • Fast optical plates ensure excellent precision in
Specifications
Specifications
1 System
Cy3™, Cy5™, FAM™, JOE™, NED™, ROX™, SYBR™ Green, TAMRA™, Texas Red™, VIC™
45cm
CCD camera, five excitation and five emission filters, halogen light source
10 to 30μL
Down to 1 copy of human RNase P gene
Throughput: 4 to 5-96-well plates per day; Scan Time: <2 hr. (Standard), 36 min. (Fast); Precision: Distinguish between 5000 to 10000 genome equivalents (two-fold copy number difference) with 99.7% confidence
49cm
9 logs of linear dynamic range
5.5°C/sec.
Fast mode: ±3.5°C/sec., Standard mode: ±1.6°C/sec.
±0.5°C