missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Quinolinecarboxaldehyde 98.0+%, TCI America™
Supplier: TCI America Q00671G
Specifications
7-Quinolinecarboxaldehyde | |
87°C | |
C10H7NO | |
1 g | |
WINWAFCAQPFBQA-UHFFFAOYSA-N | |
quinoline-7-carbaldehyde | |
4712255 | |
≥98.0% (GC) |
49573-30-0 | |
White-Yellow | |
MFCD01366121 | |
7-Formylquinoline | |
C1=CC2=C(C=C(C=C2)C=O)N=C1 | |
157.172 | |
157.17 | |
Crystalline Powder |
Chemical Identifiers
49573-30-0 | |
157.172 | |
WINWAFCAQPFBQA-UHFFFAOYSA-N | |
4712255 | |
C1=CC2=C(C=C(C=C2)C=O)N=C1 |
C10H7NO | |
MFCD01366121 | |
7-Formylquinoline | |
quinoline-7-carbaldehyde |
Safety and Handling
TSCA : No