missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Quinolinecarboxaldehyde 98.0+%, TCI America™
$428.02 - $1305.63
Chemical Identifiers
CAS | 49573-30-0 |
---|---|
Molecular Formula | C10H7NO |
Molecular Weight (g/mol) | 157.172 |
MDL Number | MFCD01366121 |
InChI Key | WINWAFCAQPFBQA-UHFFFAOYSA-N |
Synonym | 7-Formylquinoline |
PubChem CID | 4712255 |
IUPAC Name | quinoline-7-carbaldehyde |
SMILES | C1=CC2=C(C=C(C=C2)C=O)N=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
Q0067250MG
|
TCI America
Q0067250MG |
250 mg |
Each for $428.02
|
|
|||||
Q00671G
|
TCI America
Q00671G |
1 g |
Each for $1,305.63
|
|
|||||
Chemical Identifiers
49573-30-0 | |
157.172 | |
WINWAFCAQPFBQA-UHFFFAOYSA-N | |
4712255 | |
C1=CC2=C(C=C(C=C2)C=O)N=C1 |
C10H7NO | |
MFCD01366121 | |
7-Formylquinoline | |
quinoline-7-carbaldehyde |
Specifications
49573-30-0 | |
White-Yellow | |
MFCD01366121 | |
7-Formylquinoline | |
C1=CC2=C(C=C(C=C2)C=O)N=C1 | |
157.172 | |
157.17 | |
Crystalline Powder |
87°C | |
C10H7NO | |
250 mg | |
WINWAFCAQPFBQA-UHFFFAOYSA-N | |
quinoline-7-carbaldehyde | |
4712255 | |
≥98.0% (GC) | |
7-Quinolinecarboxaldehyde |
Safety and Handling
TSCA : No