missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Phenyl-1-heptanol, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1253303

 View more versions of this product

Catalog No. AAL1253303



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7-Phenyl-1-heptanol
0.962
C13H20O
MFCD00037315
1865431
UXMUSYTXSNVRMW-UHFFFAOYSA-N
7-phenylheptan-1-ol
154271
97%
3208-25-1
115°C to 117°C (0.5 mmHg)
1.508
1 g
benzeneheptanol, 7-phenyl-1-heptanol, 7-phenylheptanol, phenylheptyl alcohol, 7-phenyl-n-heptanol, acmc-20alwo, 7-phenylheptyl alcohol
C1=CC=C(C=C1)CCCCCCCO
192.302
192.3

Chemical Identifiers

3208-25-1
192.302
UXMUSYTXSNVRMW-UHFFFAOYSA-N
154271
C1=CC=C(C=C1)CCCCCCCO
C13H20O
MFCD00037315
benzeneheptanol, 7-phenyl-1-heptanol, 7-phenylheptanol, phenylheptyl alcohol, 7-phenyl-n-heptanol, acmc-20alwo, 7-phenylheptyl alcohol
7-phenylheptan-1-ol
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only