Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Pentadecanone, 98%

$140.74 - $140.74

Chemical Identifiers

Chemical Name or Material 7-Pentadecanone
CAS 6064-38-6
Molecular Formula C15H30O
Formula Weight 226.4
MDL Number MFCD00027335
Synonym n-Hexyl n-octyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2476803

  SDS
alfa aesar™
B2476803
1g Each for $140.74
Description & Specifications

Specifications

Formula Weight 226.4
Chemical Name or Material 7-Pentadecanone
Assay 98%
Molecular Formula C15H30O
MDL Number MFCD00027335
CAS 6064-38-6
Synonym n-Hexyl n-octyl ketone
Melting Point 30° to 32°C
Quantity 1g