missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Nitroindole, TCI America™

$140.56 - $377.38

Chemical Identifiers

CAS 6960-42-5
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00005683
InChI Key LZJGQIVWUKFTRD-UHFFFAOYSA-N
Synonym 7-nitroindole, 1h-indole, 7-nitro, indole, 7-nitro, 7-nitro indole, pubchem7441, 7-nitroindole, acmc-1b2jq, 5-20-07-00044 beilstein handbook reference, ksc353k7l, 1h-indole, 7-nitro-9ci
PubChem CID 23396
IUPAC Name 7-nitro-1H-indole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06851G View Documents TCI America
N06851G
1g Each for $140.56
 
N06855G View Documents TCI America
N06855G
5g Each for $377.38
 
Specifications

Chemical Identifiers

6960-42-5
162.148
LZJGQIVWUKFTRD-UHFFFAOYSA-N
23396
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2
C8H6N2O2
MFCD00005683
7-nitroindole, 1h-indole, 7-nitro, indole, 7-nitro, 7-nitro indole, pubchem7441, 7-nitroindole, acmc-1b2jq, 5-20-07-00044 beilstein handbook reference, ksc353k7l, 1h-indole, 7-nitro-9ci
7-nitro-1H-indole

Specifications

Yellow
1g
C8H6N2O2
7-nitroindole, 1h-indole, 7-nitro, indole, 7-nitro, 7-nitro indole, pubchem7441, 7-nitroindole, acmc-1b2jq, 5-20-07-00044 beilstein handbook reference, ksc353k7l, 1h-indole, 7-nitro-9ci
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2
162.148
162.15
≥98.0% (GC)
97°C
6960-42-5
MFCD00005683
LZJGQIVWUKFTRD-UHFFFAOYSA-N
7-nitro-1H-indole
23396
Crystal-Powder at 20°C
7-Nitroindole
Safety and Handling

Safety and Handling

RTECSNumber : NM1169000

TSCA : No