Please login to your online account to display your discounted pricing

7-Nitroindole, 99%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  123140010

 View more versions of this product

Catalog No. AC123140010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 162.15
Quantity 1g
Color Yellow
Beilstein 20, III, 3194
Melting Point 95° to 98°C
CAS Max % 100.0
Chemical Name or Material 7-Nitroindole
Assay Percent Range 99%
Molecular Formula C8H6N2O2
CAS 6960-42-5
Packaging Glass bottle
MDL Number MFCD00005683
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 7-Nitroindole
CAS 6960-42-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00005683
Accessories

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.

SDS & Safety Documents