Please login to your online account to display your discounted pricing

7-Nitroindole, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 7-Nitroindole
CAS 6960-42-5
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00005683
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC123140010

  SDS
thermo fisher scientific
123140010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5%
Formula Weight 162.15
Physical Form Crystalline Powder
Quantity 1g
Color Yellow
Beilstein 20, III, 3194
Melting Point 95° to 98°C
CAS Max % 100.0
Chemical Name or Material 7-Nitroindole
Molecular Formula C8H6N2O2
CAS 6960-42-5
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Suspected of causing genetic defects.
MDL Number MFCD00005683
CAS Min % 98.5
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Suspected of causing genetic defects.