Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Nitroindole, 97%

$103.07 - $1,265.68

Chemical Identifiers

Chemical Name or Material 7-Nitroindole
CAS 6960-42-5
Molecular Formula C8H6N2O2
Formula Weight 162.15
MDL Number MFCD00005683
Beilstein 130874
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1938403

  SDS
alfa aesar™
A1938403
1g Each for $103.07

AAA1938406

  SDS
alfa aesar™
A1938406
5g Each for $328.47

AAA1938414

  SDS
alfa aesar™
A1938414
25g Each for $1,265.68
Description & Specifications

Specifications

Formula Weight 162.15
Chemical Name or Material 7-Nitroindole
Assay 97%
Molecular Formula C8H6N2O2
MDL Number MFCD00005683
CAS 6960-42-5
Beilstein 130874
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H341
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number NM1169000
Odor Odorless
Melting Point 94° to 96°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H341
Suspected of causing genetic defects.