missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Nitroindole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1938414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
7-Nitroindole | |
94°C to 96°C | |
C8H6N2O2 | |
25 g | |
7-nitroindole, 1h-indole, 7-nitro, indole, 7-nitro, 7-nitro indole, pubchem7441, acmc-1b2jq, 5-20-07-00044 beilstein handbook reference, ksc353k7l, 1h-indole, 7-nitro-9ci | |
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2 | |
162.148 | |
162.15 |
6960-42-5 | |
Odorless | |
MFCD00005683 | |
130874 | |
LZJGQIVWUKFTRD-UHFFFAOYSA-N | |
7-nitro-1H-indole | |
23396 | |
97% |
Chemical Identifiers
6960-42-5 | |
162.148 | |
LZJGQIVWUKFTRD-UHFFFAOYSA-N | |
23396 | |
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2 |
C8H6N2O2 | |
MFCD00005683 | |
7-nitroindole, 1h-indole, 7-nitro, indole, 7-nitro, 7-nitro indole, pubchem7441, acmc-1b2jq, 5-20-07-00044 beilstein handbook reference, ksc353k7l, 1h-indole, 7-nitro-9ci | |
7-nitro-1H-indole |
Safety and Handling
GHS H Statement
H341
Suspected of causing genetic defects.
P201-P202-P281-P308+P313-P501c
H341
EINECSNumber : 000-000-0
RTECSNumber : NM1169000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only