missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Nitroindole-3-carboxaldehyde, 97%

$30.38 - $433.34

Chemical Identifiers

CAS 10553-14-7
Molecular Formula C9H6N2O3
Molecular Weight (g/mol) 190.158
MDL Number MFCD01074519
InChI Key ADGKBVRTGVODMM-UHFFFAOYSA-N
Synonym 7-nitroindole-3-carboxyaldehyde, 7-nitroindole-3-carboxaldehyde, 7-nitroindole-3-carbaldehyde, 3-carboxy-7-nitro-1h-indole, 7-nitro-1h-indole-3-carboxaldehyde, 7-nitroindole-3-aldehyde, pubchem9125, 3-formyl-7-nitro-1h-indole, 7-nitroindole 3-carboxaldehyde, 7-nitroindole-3-carboxaldehyde
PubChem CID 2762438
IUPAC Name 7-nitro-1H-indole-3-carbaldehyde
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63778MD View Documents Alfa Aesar™
H63778MD
250mg Each for $30.38
 
AAH6377803 View Documents Alfa Aesar™
H6377803
1g Each for $95.46
 
AAH6377806 View Documents Alfa Aesar™
H6377806
5g Each for $433.34
 
Specifications

Chemical Identifiers

10553-14-7
190.158
ADGKBVRTGVODMM-UHFFFAOYSA-N
2762438
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2C=O
C9H6N2O3
MFCD01074519
7-nitroindole-3-carboxyaldehyde, 7-nitroindole-3-carboxaldehyde, 7-nitroindole-3-carbaldehyde, 3-carboxy-7-nitro-1h-indole, 7-nitro-1h-indole-3-carboxaldehyde, 7-nitroindole-3-aldehyde, pubchem9125, 3-formyl-7-nitro-1h-indole, 7-nitroindole 3-carboxaldehyde, 7-nitroindole-3-carboxaldehyde
7-nitro-1H-indole-3-carbaldehyde

Specifications

250mg
C9H6N2O3
7-nitroindole-3-carboxyaldehyde, 7-nitroindole-3-carboxaldehyde, 7-nitroindole-3-carbaldehyde, 3-carboxy-7-nitro-1h-indole, 7-nitro-1h-indole-3-carboxaldehyde, 7-nitroindole-3-aldehyde, pubchem9125, 3-formyl-7-nitro-1h-indole, 7-nitroindole 3-carboxaldehyde, 7-nitroindole-3-carboxaldehyde
C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2C=O
190.158
190.16
97%
10553-14-7
MFCD01074519
ADGKBVRTGVODMM-UHFFFAOYSA-N
7-nitro-1H-indole-3-carbaldehyde
2762438
Solid
7-Nitroindole-3-carboxaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C