missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Nitro-3,4-dihydro-2H-1,4-benzoxazine, 97%

Manufacturer:  Alfa Aesar™ H61032MD

 View more versions of this product

Catalog No. AAH61032MD



Description

Description

Solubility
Sparingly soluble in water (0.76 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

7-Nitro-3,4-dihydro-2H-1,4-benzoxazine
120711-81-1
MFCD11603433
Sparingly soluble in water (0.76g/L at 25°C).
C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]
180.163
180.16
250mg
C8H8N2O3
2,3-dihydro-7-nitro-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine, 7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine
YKCFDUNYLMTXFC-UHFFFAOYSA-N
7-nitro-3,4-dihydro-2H-1,4-benzoxazine
18416151
97%

Chemical Identifiers

120711-81-1
180.163
YKCFDUNYLMTXFC-UHFFFAOYSA-N
18416151
C1COC2=C(N1)C=CC(=C2)[N+](=O)[O-]
C8H8N2O3
MFCD11603433
2,3-dihydro-7-nitro-1,4-benzoxazine, 2h-1,4-benzoxazine, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-2h-1,4-benzoxazine, 3,4-dihydro-7-nitro-2h-benzo b 1,4 oxazine, 7-nitro-2,3-dihydro-4h-1,4-benzoxazine, 7-nitro-2h,3h,4h-benzo e 1,4-oxazaperhydroine, 7-nitro-3,4-dihydro-2h-1,4-benzooxazine, 7-nitro-3,4-dihydro-2h-benzo 1,4 oxazine, 7-nitro-3,4-dihydro-2h-benzo b 1,4 oxazine
7-nitro-3,4-dihydro-2H-1,4-benzoxazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents