missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Nitro-3,4-dihydro-2H-1,4-benzothiazine, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H61048MD

 View more versions of this product

Catalog No. AAH61048MD



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

7-Nitro-3,4-dihydro-2H-1,4-benzothiazine
C8H8N2O2S
250 mg
LCOAYUPAESYAHF-UHFFFAOYSA-N
7-nitro-3,4-dihydro-2H-1,4-benzothiazine
20496121
97%
69373-37-1
MFCD23379852
7-nitro-3,4-dihydro-2h-benzo b 1,4 thiazine
[O-][N+](=O)C1=CC2=C(NCCS2)C=C1
196.22
196.22

Chemical Identifiers

69373-37-1
196.22
LCOAYUPAESYAHF-UHFFFAOYSA-N
20496121
[O-][N+](=O)C1=CC2=C(NCCS2)C=C1
C8H8N2O2S
MFCD23379852
7-nitro-3,4-dihydro-2h-benzo b 1,4 thiazine
7-nitro-3,4-dihydro-2H-1,4-benzothiazine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only