missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Nitro-2H-1,4-benzothiazin-3(4H)-one, 97%

$49.20 - $155.76

Chemical Identifiers

CAS 21762-75-4
Molecular Formula C8H6N2O3S
Molecular Weight (g/mol) 210.207
MDL Number MFCD23379853
InChI Key OOICZQBVVODNDJ-UHFFFAOYSA-N
Synonym 7-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 7-nitro-3,4-dihydro-3-oxo-2h 1,4 benzothiazine, 7-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 7-nitro-4h-benzo 1,4 thiazin-3-one, 7-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 7-nitro, 3,4-dihydro-7-nitro-2h-1,4-benzothiazin-3-one, 7-hydroxy oxido amino-2h-1,4-benzothiazin-3 4h-one
PubChem CID 370121
IUPAC Name 7-nitro-4H-1,4-benzothiazin-3-one
SMILES C1C(=O)NC2=C(S1)C=C(C=C2)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH61050MD View Documents Alfa Aesar™
H61050MD
250mg Each for $49.20
 
AAH6105003 View Documents Alfa Aesar™
H6105003
1g Each for $155.76
 
Specifications

Chemical Identifiers

21762-75-4
210.207
OOICZQBVVODNDJ-UHFFFAOYSA-N
370121
C1C(=O)NC2=C(S1)C=C(C=C2)[N+](=O)[O-]
C8H6N2O3S
MFCD23379853
7-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 7-nitro-3,4-dihydro-3-oxo-2h 1,4 benzothiazine, 7-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 7-nitro-4h-benzo 1,4 thiazin-3-one, 7-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 7-nitro, 3,4-dihydro-7-nitro-2h-1,4-benzothiazin-3-one, 7-hydroxy oxido amino-2h-1,4-benzothiazin-3 4h-one
7-nitro-4H-1,4-benzothiazin-3-one

Specifications

250mg
C8H6N2O3S
7-nitro-2h-benzo b 1,4 thiazin-3 4h-one, 7-nitro-3,4-dihydro-3-oxo-2h 1,4 benzothiazine, 7-nitro-2,4-dihydro-1,4-benzothiazin-3-one, 7-nitro-4h-benzo 1,4 thiazin-3-one, 7-nitro-2h-1,4-benzothiazin-3 4h-one, 2h-1,4-benzothiazin-3 4h-one, 7-nitro, 3,4-dihydro-7-nitro-2h-1,4-benzothiazin-3-one, 7-hydroxy oxido amino-2h-1,4-benzothiazin-3 4h-one
C1C(=O)NC2=C(S1)C=C(C=C2)[N+](=O)[O-]
210.207
210.21
7-Nitro-2H-1,4-benzothiazin-3(4H)-one
21762-75-4
MFCD23379853
OOICZQBVVODNDJ-UHFFFAOYSA-N
7-nitro-4H-1,4-benzothiazin-3-one
370121
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning