missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Nitro-1-tetralone, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0185309
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
7-Nitro-1-tetralone | |
103°C to 108°C | |
MFCD00019661 | |
1570515 | |
GWAQYWSNCVEJMW-UHFFFAOYSA-N | |
7-nitro-3,4-dihydro-2H-naphthalen-1-one | |
38445 | |
97% |
40353-34-2 | |
C10H9NO3 | |
10 g | |
7-nitro-1-tetralone, 7-nitrotetralone, 7-nitro-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-1 2h-naphthalenone, 3,4-dihydro-7-nitronaphthalen-1 2h-one, wln: l66 bvt&j inw, 7-nitro-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone,4-dihydro-7-nitro, 7-nitro-tetralone | |
C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1 | |
191.186 | |
191.19 |
Chemical Identifiers
40353-34-2 | |
191.186 | |
GWAQYWSNCVEJMW-UHFFFAOYSA-N | |
38445 | |
C1CC2=C(C=C(C=C2)[N+](=O)[O-])C(=O)C1 |
C10H9NO3 | |
MFCD00019661 | |
7-nitro-1-tetralone, 7-nitrotetralone, 7-nitro-3,4-dihydronaphthalen-1 2h-one, 1 2h-naphthalenone, 3,4-dihydro-7-nitro, 3,4-dihydro-7-nitro-1 2h-naphthalenone, 3,4-dihydro-7-nitronaphthalen-1 2h-one, wln: l66 bvt&j inw, 7-nitro-2,3,4-trihydronaphthalen-1-one, 1 2h-naphthalenone,4-dihydro-7-nitro, 7-nitro-tetralone | |
7-nitro-3,4-dihydro-2H-naphthalen-1-one |
Safety and Handling
EINECSNumber : 254-887-6
RTECSNumber : QK5025000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only