Please login to your online account to display your discounted pricing

7-NINA, Tocris


Non-selective NOS inhibitor; Sodium salt of 7-nitroindazole

Quantity 10mg

Manufacturer: r & d systems  080010

 View more versions of this product

Catalog No. 080010

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 185.12
Synonym 7-Nitroindazole monosodium salt
Assay Percent Range >98%
Chemical Name or Material 7-NINA
Molecular Formula C7H4N3O2Na
Recommended Storage Desiccate at -20°C
CAS 161467-34-1

Specifications

Chemical Name or Material 7-NINA
Assay Percent Range >98%
Molecular Formula C7H4N3O2Na
Formula Weight 185.12
Synonym 7-Nitroindazole monosodium salt
Accessories

Desiccate at -20°C