Please login to your online account to display your discounted pricing

7-NINA, Tocris

Non-selective NOS inhibitor; Sodium salt of 7-nitroindazole

$108.46 - $458.10

Chemical Identifiers

Chemical Name or Material 7-NINA
Assay Percent Range >98%
Molecular Formula C7H4N3O2Na
Formula Weight 185.12
Synonym 7-Nitroindazole monosodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

080010

 
r & d systems
080010
10mg Each for $108.46

080050

 
r & d systems
080050
50mg Each for $458.10
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 185.12
Chemical Name or Material 7-NINA
Assay Percent Range >98%
Molecular Formula C7H4N3O2Na
Synonym 7-Nitroindazole monosodium salt
Recommended Storage Desiccate at -20°C
CAS 161467-34-1
Safety and Handling

Desiccate at -20°C