missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Methylquinoline (contains 25% 5-form at maximum) 75.0+%, TCI America™
Supplier: TCI America M041725G
Specifications
7-Methylquinoline (contains 25% 5-form at maximum) | |
Yellow | |
MFCD00006805 | |
quinoline, 7-methyl, m-toluquinoline, unii-eow995k43c, 7-methyl quinoline, ccris 2895, quinolinol,7-methyl, 7-methyl-quinoline, 7-methylchinolin, 7-methylchinolin;, pubchem5811 | |
CC1=CC2=C(C=CC=N2)C=C1 | |
143.189 | |
143.19 | |
Liquid |
612-60-2 | |
C10H9N | |
25 g | |
KDYVCOSVYOSHOL-UHFFFAOYSA-N | |
7-methylquinoline | |
11927 | |
≥75.0% (GC) |
Chemical Identifiers
612-60-2 | |
143.189 | |
KDYVCOSVYOSHOL-UHFFFAOYSA-N | |
11927 | |
CC1=CC2=C(C=CC=N2)C=C1 |
C10H9N | |
MFCD00006805 | |
quinoline, 7-methyl, m-toluquinoline, unii-eow995k43c, 7-methyl quinoline, ccris 2895, quinolinol,7-methyl, 7-methyl-quinoline, 7-methylchinolin, 7-methylchinolin;, pubchem5811 | |
7-methylquinoline |
Safety and Handling
EINECSNumber : (5)-0796
RTECSNumber : VC0560000
TSCA : Yes