Please login to your online account to display your discounted pricing

7-Methylbenzo[d]isoxazol-3-ol, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 7-Methylbenzo[d]isoxazol-3-ol
CAS 36238-83-2
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H7NO2
Formula Weight 149.15
MDL Number MFCD00104669
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

RF03630DA

 
fisher scientific company
RF03630DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

RF03630EA

 
fisher scientific company
RF03630EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 149.15
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 7-Methylbenzo[d]isoxazol-3-ol
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 36238-83-2
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Molecular Formula C8H7NO2
MDL Number MFCD00104669
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.