missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methyl-4-indanyl acetate, 97%

$85.51 - $323.04

Chemical Identifiers

CAS 175136-12-6
Molecular Formula C12H14O2
Molecular Weight (g/mol) 190.242
MDL Number MFCD00173682
InChI Key MABWFAYFFNHAOF-UHFFFAOYSA-N
Synonym 1h-inden-4-ol,2,3-dihydro-7-methyl-, 4-acetate, 1h-inden-4-ol,2,3-dihydro-7-methyl-,4-acetate, 4-acetoxy-7-methylindane, 7-methyl-2,3-dihydro-1h-inden-4-yl acetate, 7-methylindan-4-yl acetate, acmc-1c5pj
PubChem CID 2775024
IUPAC Name (7-methyl-2,3-dihydro-1H-inden-4-yl) acetate
SMILES CC1=C2CCCC2=C(C=C1)OC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2092303
View Documents
Alfa Aesar™
B2092303
1g Each for $85.51
 
AAB2092306
View Documents
Alfa Aesar™
B2092306
5g Each for $323.04
 
Specifications

Chemical Identifiers

175136-12-6
190.242
MABWFAYFFNHAOF-UHFFFAOYSA-N
2775024
CC1=C2CCCC2=C(C=C1)OC(=O)C
C12H14O2
MFCD00173682
1h-inden-4-ol,2,3-dihydro-7-methyl-, 4-acetate, 1h-inden-4-ol,2,3-dihydro-7-methyl-,4-acetate, 4-acetoxy-7-methylindane, 7-methyl-2,3-dihydro-1h-inden-4-yl acetate, 7-methylindan-4-yl acetate, acmc-1c5pj
(7-methyl-2,3-dihydro-1H-inden-4-yl) acetate

Specifications

113°C to 114°C (0.7mmHg)
175136-12-6
MFCD00173682
MABWFAYFFNHAOF-UHFFFAOYSA-N
(7-methyl-2,3-dihydro-1H-inden-4-yl) acetate
2775024
97%
1g
C12H14O2
1h-inden-4-ol,2,3-dihydro-7-methyl-, 4-acetate, 1h-inden-4-ol,2,3-dihydro-7-methyl-,4-acetate, 4-acetoxy-7-methylindane, 7-methyl-2,3-dihydro-1h-inden-4-yl acetate, 7-methylindan-4-yl acetate, acmc-1c5pj
CC1=C2CCCC2=C(C=C1)OC(=O)C
190.242
190.24
7-Methyl-4-indanyl acetate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures