Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methyl-4-indanol, 97%

$114.36 - $315.55

Chemical Identifiers

Chemical Name or Material 7-Methyl-4-indanol
CAS 16400-13-8
Molecular Formula C10H12O
Formula Weight 148.21
MDL Number MFCD00173675
Synonym 4-Hydroxy-7-methylindane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2034309

 
alfa aesar™
B2034309
10g Each for $315.55

AAB2034388

 
alfa aesar™
B2034388
2.5g Each for $114.36
Description & Specifications

Specifications

Formula Weight 148.21
Chemical Name or Material 7-Methyl-4-indanol
Assay 97%
Molecular Formula C10H12O
MDL Number MFCD00173675
CAS 16400-13-8
Melting Point 80° to 82°C
Quantity 10g
Synonym 4-Hydroxy-7-methylindane
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
7-Methyl-4-indanol is used as an intermediate in synthetic chemistry.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.