missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Methyl-1,5-benzodioxepan-3-one, TCI America™

$232.23 - $722.66

Chemical Identifiers

CAS 28940-11-6
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD07371373
InChI Key SWUIQEBPZIHZQS-UHFFFAOYSA-N
PubChem CID 120101
IUPAC Name 7-methyl-1,5-benzodioxepin-3-one
SMILES CC1=CC2=C(C=C1)OCC(=O)CO2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M204525G View Documents TCI America
M204525G
25g Each for $722.66
 
M20455G View Documents TCI America
M20455G
5g Each for $232.23
 
Specifications

Chemical Identifiers

28940-11-6
178.187
SWUIQEBPZIHZQS-UHFFFAOYSA-N
7-methyl-1,5-benzodioxepin-3-one
C10H10O3
MFCD07371373
120101
CC1=CC2=C(C=C1)OCC(=O)CO2

Specifications

91°C
39°C
28940-11-6
MFCD07371373
CC1=CC2=C(C=C1)OCC(=O)CO2
178.187
178.19
≥98.0% (GC)
White-Yellow
25g
C10H10O3
SWUIQEBPZIHZQS-UHFFFAOYSA-N
7-methyl-1,5-benzodioxepin-3-one
120101
Crystal-Powder at 20°C
7-Methyl-1,5-benzodioxepan-3-one
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator