missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

7-Methyl-1,5-benzodioxepan-3-one 98.0+%, TCI America™

Manufacturer:  Tci America M20455G

 View more versions of this product

Catalog No. M20455G



Specifications

Specifications

7-Methyl-1,5-benzodioxepan-3-one
White-Yellow
5g
C10H10O3
SWUIQEBPZIHZQS-UHFFFAOYSA-N
7-methyl-1,5-benzodioxepin-3-one
120101
Crystal-Powder at 20°C
91°C
39°C
28940-11-6
MFCD07371373
CC1=CC2=C(C=C1)OCC(=O)CO2
178.187
178.19
≥98.0% (GC)

Chemical Identifiers

28940-11-6
178.187
SWUIQEBPZIHZQS-UHFFFAOYSA-N
7-methyl-1,5-benzodioxepin-3-one
C10H10O3
MFCD07371373
120101
CC1=CC2=C(C=C1)OCC(=O)CO2
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Refrigerator

Documents
Promotions

Promotions