missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
7-Methyl-1,5-benzodioxepan-3-one 98.0+%, TCI America™
Supplier: TCI America M204525G
Specifications
7-Methyl-1,5-benzodioxepan-3-one | |
39°C | |
91°C | |
MFCD07371373 | |
SWUIQEBPZIHZQS-UHFFFAOYSA-N | |
7-methyl-1,5-benzodioxepin-3-one | |
120101 | |
≥98.0% (GC) |
28940-11-6 | |
White-Yellow | |
C10H10O3 | |
25 g | |
CC1=CC2=C(C=C1)OCC(=O)CO2 | |
178.187 | |
178.19 | |
Crystalline Powder |
Chemical Identifiers
28940-11-6 | |
178.187 | |
SWUIQEBPZIHZQS-UHFFFAOYSA-N | |
7-methyl-1,5-benzodioxepin-3-one |
C10H10O3 | |
MFCD07371373 | |
120101 | |
CC1=CC2=C(C=C1)OCC(=O)CO2 |
Safety and Handling
TSCA : Yes
Recommended Storage : Refrigerator