Alfa Aesar™ 7-Methoxyindole, 97%
Manufacturer:  Alfa Aesar™ H6376706
 View more versions of this product
Catalog No. AAH6376706
Specifications
Specifications
7-Methoxyindole
1.612
5g
C9H9NO
7-methoxyindole, 7-methoxy indole, 1h-indole, 7-methoxy, indole, 7-methoxy, methoxyindole 7-, unii-9r05qk9rp4, 7-methoxy-indole, pubchem7434, acmc-209hpi, 7-methoxyindole
COC1=CC=CC2=C1NC=C2
147.177
147.17
97%
1.126g/mL
108°C to 110°C (0.3mmHg)
3189-22-8
MFCD00047203
FSOPPXYMWZOKRM-UHFFFAOYSA-N
7-methoxy-1H-indole
76660
Solid
Chemical Identifiers
3189-22-8
147.177
FSOPPXYMWZOKRM-UHFFFAOYSA-N
76660
COC1=CC=CC2=C1NC=C2
C9H9NO
MFCD00047203
7-methoxyindole, 7-methoxy indole, 1h-indole, 7-methoxy, indole, 7-methoxy, methoxyindole 7-, unii-9r05qk9rp4, 7-methoxy-indole, pubchem7434, acmc-209hpi, 7-methoxyindole
7-methoxy-1H-indole
SDS Documents