Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxyflavone, 99%

$88.82 - $1,349.95

Chemical Identifiers

Chemical Name or Material 7-Methoxyflavone
CAS 22395-22-8
Molecular Formula C16H12O3
Formula Weight 252.27
MDL Number MFCD00017462
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2485003

 
alfa aesar™
B2485003
1g Each for $88.82

AAB2485006

 
alfa aesar™
B2485006
5g Each for $290.92

AAB2485014

 
alfa aesar™
B2485014
25g Each for $1,349.95
Description & Specifications

Specifications

Formula Weight 252.27
Chemical Name or Material 7-Methoxyflavone
Assay 99%
Molecular Formula C16H12O3
MDL Number MFCD00017462
CAS 22395-22-8
Melting Point 108° to 112°C
Quantity 1g