Please login to your online account to display your discounted pricing

Alfa Aesar™ 7-Methoxyflavanone, 98%

$67.66 - $197.39

Chemical Identifiers

Chemical Name or Material 7-Methoxyflavanone
CAS 21785-09-1
Molecular Formula C16H14O3
Formula Weight 254.29
MDL Number MFCD00017486
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2535803

  SDS
alfa aesar™
B2535803
1g Each for $67.66

AAB2535806

  SDS
alfa aesar™
B2535806
5g Each for $197.39
Description & Specifications

Specifications

Formula Weight 254.29
Health Hazard 1 Warning
Molecular Formula C16H14O3
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00017486
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 86° to 90°C
CAS 21785-09-1
Chemical Name or Material 7-Methoxyflavanone
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.